Learn More
ATCC Vibrio campbellii; BB120
Catalog No. 502384681
Encompass
Vibrio campbellii strain BB120 is a whole-genome sequenced bacterium isolated from a marine source. It is one of several strains (ATCC BAA-1116 through ATCC BAA-1121) included in a "quorum sensing reporter kit" (ATCC MP-6) developed by the depositor. This wild type strain responds to both AI-1 and AI-2 autoinducers.
Catalog No. 50-238-4681 Supplier ATCC Supplier No. BAA1116
Vibrio campbellii strain BB120 is a whole-genome sequenced bacterium isolated from a marine source. It is one of several strains (ATCC BAA-1116 through ATCC BAA-1121) included in a "quorum sensing reporter kit" (ATCC MP-6) developed by the depositor. This wild type strain responds to both AI-1 and AI-2 autoinducers.