Learn More
ATCC Vibrio campbellii; BB721
Catalog No. 502383217
Encompass
Vibrio campbellii strain BB721 is a mutant derived from Vibrio campbellii strain BB120 (ATCC BAA-1116). This strain produces light; the expression of luminescence is not influenced by culture density.
Catalog No. 50-238-3217 Supplier ATCC Supplier No. 700106
Vibrio campbellii strain BB721 is a mutant derived from Vibrio campbellii strain BB120 (ATCC BAA-1116). This strain produces light; the expression of luminescence is not influenced by culture density.