Learn More
ATCC Vibrio campbellii; Strain MM30
Supplier:  ATCC BAA1120
Encompass
Vibrio campbellii strain MM30 is a whole-genome sequenced bacterium that was derived from Vibrio campbellii strain BB120 (ATCC BAA-1116). This culture is one of several strains included in a “quorum sensing kit” (ATCC MP-6) developed by the depositor. It is a mutant sensor strain that responds only to the AI-1 autoinducer.
Catalog No. 50-237-9846