Learn More
Microbiologics™ Vibrio furnissii NCTC 11218
SDP
Catalog No. 23017212 Shop All Microbiologics Products
Change view
Click to view available options
Product Type:
Lyfo-Disk QC Control
Quality Control Organism
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
23-017-212 Lyfo-Disk QC Control
23-017-211 Quality Control Organism
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-017-212 Supplier Microbiologics™ Supplier No. 0720L
Add to Cart
Add to Cart
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single strain of Vibrio furnissii in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in pack of 2
  • NCTC 11218
Specifications
ATCC® ATCC 11218
Biosafety Level 1
Product Type Lyfo-Disk QC Control
Quantity 6/Pk.
Species Vibrio furnissii
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.