Learn More
ATCC Vibrio parahaemolyticus; EB 101
Supplier:  ATCC 17802
Encompass
Vibrio parahaemolyticus strain EB101 was isolated from a case of Shirasu food poisoning in Japan. This type strain is whole-genome sequenced and has applications as a control strain for the detection of Vibrio parahaemolyticus and can be used in enteric research and waterborne pathogen research.
Catalog No. 50-238-2532
 Restricted Item