Learn More
ATCC Vibrio parahaemolyticus; Strain EB 101; genomic DNA
Catalog No. 502382534
Encompass
Genomic DNA isolated from Vibrio parahaemolyticus strain EB 101 (ATCC 17802). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-2534 Supplier ATCC Supplier No. 17802D5
Genomic DNA isolated from Vibrio parahaemolyticus strain EB 101 (ATCC 17802). This whole-genome sequenced product can be used in PCR and other molecular biology applications.