Learn More
ATCC Vibrio vulnificus
Catalog No. 502381683
Encompass
Vibrio vulnificus strain 324 was isolated from human blood in Florida. This whole-genome sequenced product produces hemolysins and has applications in waterborne pathogen research and as a control strain for the detection of Vibrio vulnificus.
Catalog No. 50-238-1683 Supplier ATCC Supplier No. 27562
Vibrio vulnificus strain 324 was isolated from human blood in Florida. This whole-genome sequenced product produces hemolysins and has applications in waterborne pathogen research and as a control strain for the detection of Vibrio vulnificus.