Learn More
ATCC Rhizobium species
Catalog No. 502381319
Encompass
Previously known as Rhizobium radiobacter. This bacterium has been used as a biocontrol agent to protect plants from crown gall disease for more than 25 years.
Catalog No. 50-238-1319 Supplier ATCC Supplier No. BAA868
Previously known as Rhizobium radiobacter. This bacterium has been used as a biocontrol agent to protect plants from crown gall disease for more than 25 years.