Learn More
Medchemexpress LLC Vic phosphoramidite, 6-isomer | 1414265-81-8 | 98.5% | 1023.37 g/mol | C52H59Cl3N3O10P | 10 MG
SDP
Catalog No. 5000297191
Encompass_Preferred
VIC phosphoramidite, 6-isomer is a fluorescent phosphoramidite reagent used to incorporate the VIC dye into oligonucleotides (commonly at the 5'-terminus) or to conjugate VIC to other molecules for fluorescence labeling and detection in molecular biology assays.
  • Facilitates 5'-end labeling of oligonucleotides.
  • High purity (98.5%).
  • White to off-white solid, suitable for standard handling.
  • Stable when protected from light; short-term storage at -20°C, long-term at -80°C in solvent.
  • Available in small research pack sizes for oligonucleotide synthesis workflows.
Catalog No. 50-002-97191 Supplier Medchemexpress LLC Supplier No. HYD189910MG
VIC phosphoramidite, 6-isomer is a fluorescent phosphoramidite reagent used to incorporate the VIC dye into oligonucleotides (commonly at the 5'-terminus) or to conjugate VIC to other molecules for fluorescence labeling and detection in molecular biology assays.
  • Facilitates 5'-end labeling of oligonucleotides.
  • High purity (98.5%).
  • White to off-white solid, suitable for standard handling.
  • Stable when protected from light; short-term storage at -20°C, long-term at -80°C in solvent.
  • Available in small research pack sizes for oligonucleotide synthesis workflows.