Learn More
Marketlab VIC Video Fluoroscopy Imaging Chair
Description
- Radiolucent, removable back section provides a clean, unobstructed site for imaging
- Backrest and leg sections adjust from 90° upright to full supine and 10° Trendelenburg positions
- Four-way stretch urethane-covered HD foam surface is impervious to fluid
- Improves turn-around-time and streamlines patient handling
- Easy-to-maneuver shortened base
- 325 lb. weight capacity
Specifications
Specifications
| Arm Style | Standard |
| For Use With (Application) | Fluoroscopy |