Learn More
Medchemexpress LLC HY-118284 10mg Medchemexpress, Vicagrel CAS:1314081-53-2 Purity:>98%
SDP
Catalog No. 501969502
Encompass_Preferred
Medchemexpress, HY-118284 10mg Vicagrel CAS:1314081-53-2 Vicagrel, an acetate derivative of Clopidogrel, is a P2Y12 platelet inhibitor potentially for the treatment of thrombosis, the substrate of carboxylesterase 2 (CES2). Vicagrel demonstrates a favorable safety profile and excellent anti-platelet activity, which could be an anti-platelet drug and for the unmet medical needs of cardiovascular diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9502 Supplier Medchemexpress LLC Supplier No. HY11828410MG
Medchemexpress, HY-118284 10mg Vicagrel CAS:1314081-53-2 Vicagrel, an acetate derivative of Clopidogrel, is a P2Y12 platelet inhibitor potentially for the treatment of thrombosis, the substrate of carboxylesterase 2 (CES2). Vicagrel demonstrates a favorable safety profile and excellent anti-platelet activity, which could be an anti-platelet drug and for the unmet medical needs of cardiovascular diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.