Learn More
Beckman Coulter Beckman Coulter Vi-CELL MetaFLEX Bioanalyte Analyzer
Supplier:  Beckman Coulter B93015
Encompass
The Vi-CELL MetaFLEX is a benchtop bioanalyte analyzer for rapid, low-volume metabolic and dissolved gas measurements in cell culture and bioprocess applications. It provides automated quality control and data logging to support regulated workflows.
  • Performs pH, pO2, pCO2, glucose, lactate, and electrolyte measurements.
  • Requires sample volumes as low as 65 μL.
  • Delivers results in about 35 seconds per sample.
  • Includes three on-board QC solutions and customizable QC scheduling.
  • Provides security and data logging to support 21 CFR Part 11 compliance.
Catalog No. NC2315780
Request a Quote
To request a quote for this item, please select Request a Quote to reach our dedicated support team.