Learn More
TARGETMOL CHEMICALS INC VICENIN 2 5MG
SDP
Catalog No. 502576866
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vicenin 2 is an angiotensin-converting enzyme (ACE) inhibitor (IC50=43.83 uM) from the aerial parts of Desmodium styracifolium. Vicenin 2 is an inhibitor of (alpha)-glucosidase PTP1B and RLAR. Vicenin 2 has hepatoprotective anti-cancer antioxidant and anti-inflammatory activities and DTL co-administration is more effective than either of the single agents in androgen-independent prostate cancer. Vicenin 2 might be a useful lead for the development of multiple target-oriented therapeutic modalities for the treatment of diabetes and diabetes-associated complications. Vicenin 2 could act as a UV light barrier to protect the plants. purity: 99%
Catalog No. 50-257-6866 Supplier TARGETMOL CHEMICALS INC Supplier No. T38515MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vicenin 2 is an angiotensin-converting enzyme (ACE) inhibitor (IC50=43.83 uM) from the aerial parts of Desmodium styracifolium. Vicenin 2 is an inhibitor of (alpha)-glucosidase PTP1B and RLAR. Vicenin 2 has hepatoprotective anti-cancer antioxidant and anti-inflammatory activities and DTL co-administration is more effective than either of the single agents in androgen-independent prostate cancer. Vicenin 2 might be a useful lead for the development of multiple target-oriented therapeutic modalities for the treatment of diabetes and diabetes-associated complications. Vicenin 2 could act as a UV light barrier to protect the plants. purity: 99%