Learn More
Vici Precision Sampling, Inc. VICI-Cap GL45 3 Ports. One port is suitable for 1/16in tubing.
Catalog No. NC1675339
Encompass_Preferred
GL45 caps seal to the bottle with an O-ring (EPDM) which is resistant to commonly used HPLC solvents. If improved chemical resistance is required (e.g. for halogenated hydrocarbons), order an FEP-coated O-ring separately. One port is suitable for 1/16in tubing.
Catalog No. NC1675339 Supplier Vici Precision Sampling, Inc. Supplier No. JRS11002
GL45 caps seal to the bottle with an O-ring (EPDM) which is resistant to commonly used HPLC solvents. If improved chemical resistance is required (e.g. for halogenated hydrocarbons), order an FEP-coated O-ring separately. One port is suitable for 1/16in tubing.