Learn More
Selleck Chemical LLC Vicriviroc Malate S2004-25mg
SDP
Catalog No. 503109234
Encompass_Preferred
Vicriviroc is a potent CCR5 antagonist with IC50 of 0 91 nM showing broad-spectrum activity against genetically diverse HIV-1 isolates and also drug-resistant viruses with RTI PRI or MDR phenotypes Phase 3
Catalog No. 50-310-9234 Supplier Selleck Chemical LLC Supplier No. S200425MG
Vicriviroc is a potent CCR5 antagonist with IC50 of 0 91 nM showing broad-spectrum activity against genetically diverse HIV-1 isolates and also drug-resistant viruses with RTI PRI or MDR phenotypes Phase 3