Learn More
Selleck Chemical LLC Vicriviroc Malate S2004-5mg
SDP
Catalog No. 503109720
Encompass_Preferred
Vicriviroc is a potent CCR5 antagonist with IC50 of 0 91 nM showing broad-spectrum activity against genetically diverse HIV-1 isolates and also drug-resistant viruses with RTI PRI or MDR phenotypes Phase 3
Catalog No. 50-310-9720 Supplier Selleck Chemical LLC Supplier No. S20045MG
Vicriviroc is a potent CCR5 antagonist with IC50 of 0 91 nM showing broad-spectrum activity against genetically diverse HIV-1 isolates and also drug-resistant viruses with RTI PRI or MDR phenotypes Phase 3