Learn More
Selleck Chemical LLC Vicriviroc maleate S3494-25mg
SDP
Catalog No. 503110145
Encompass_Preferred
Vicriviroc maleate(SCH-417690 maleate SCH-D maleate) is a potent selective oral bioavailable and CNS penetrated antagonist of CCR5 with a Ki of 2 5 nM and also inhibits HIV-1 in PBMC cells
Catalog No. 50-311-0145 Supplier Selleck Chemical LLC Supplier No. S349425MG
Vicriviroc maleate(SCH-417690 maleate SCH-D maleate) is a potent selective oral bioavailable and CNS penetrated antagonist of CCR5 with a Ki of 2 5 nM and also inhibits HIV-1 in PBMC cells