Learn More
Selleck Chemical LLC Vicriviroc maleate S3494-5mg
SDP
Catalog No. 503109838
Encompass_Preferred
Vicriviroc maleate(SCH-417690 maleate SCH-D maleate) is a potent selective oral bioavailable and CNS penetrated antagonist of CCR5 with a Ki of 2 5 nM and also inhibits HIV-1 in PBMC cells
Catalog No. 50-310-9838 Supplier Selleck Chemical LLC Supplier No. S34945MG
Vicriviroc maleate(SCH-417690 maleate SCH-D maleate) is a potent selective oral bioavailable and CNS penetrated antagonist of CCR5 with a Ki of 2 5 nM and also inhibits HIV-1 in PBMC cells