Learn More
Medchemexpress LLC Vicriviroc maleate | 599179-03-0 | 98.5% | 649.70 | 5 MG
SDP
Catalog No. 5000393331
Encompass_Preferred
Vicriviroc maleate is a potent, selective, and orally bioavailable CCR5 antagonist. It inhibits HIV-1 in PBMC cells and demonstrates good oral availability in rats and monkeys, making it suitable for various research applications.
  • Potent, selective, and orally bioavailable CCR5 antagonist with a Ki of 2.5 nM
  • Inhibits HIV-1 in PBMC cells with IC90s ranging from 1.8 nM to 10 nM
  • Exhibits good oral availability in rats and monkeys (10 mg/kg)
  • Weak activity against hERG (IC50, 5.8 μM)
  • Suppresses RANTES-induced signaling with a mean IC50 of 4.2 ± 1.3 nM
  • Potently suppresses various viral isolates with geometric mean EC50s of 0.04-2.3 nM and IC90s of 0.45-18 nM
Catalog No. 50-003-93331 Supplier Medchemexpress LLC Supplier No. 5MGHY17377
Add to Cart
Add to Cart
Vicriviroc maleate is a potent, selective, and orally bioavailable CCR5 antagonist. It inhibits HIV-1 in PBMC cells and demonstrates good oral availability in rats and monkeys, making it suitable for various research applications.
  • Potent, selective, and orally bioavailable CCR5 antagonist with a Ki of 2.5 nM
  • Inhibits HIV-1 in PBMC cells with IC90s ranging from 1.8 nM to 10 nM
  • Exhibits good oral availability in rats and monkeys (10 mg/kg)
  • Weak activity against hERG (IC50, 5.8 μM)
  • Suppresses RANTES-induced signaling with a mean IC50 of 4.2 ± 1.3 nM
  • Potently suppresses various viral isolates with geometric mean EC50s of 0.04-2.3 nM and IC90s of 0.45-18 nM