Learn More
Medchemexpress LLC Vicriviroc maleate | 599179-03-0 | 99.9% | 649.70 | 100 MG
SDP
Catalog No. 5000384999
Encompass_Preferred
Vicriviroc maleate is a potent, selective, oral bioavailable and CNS penetrated antagonist of CCR5, with a Ki of 2.5 nM. It also inhibits HIV-1 in PBMC cells, with IC90s of 3.3 nM (JrFL), 2.8 nM (ADA-M), 1.8 nM (301657), 4.9 nM (JV1083) and 10 nM (RU 570).
  • Potent, selective, and oral bioavailable inhibitor of CCR5.
  • Inhibits HIV-1 in PBMC cells.
  • Demonstrates a mean IC50 and IC90 of 0.45 nM and 4 nM for a panel of HIV isolates.
  • Exhibits good oral availability in rats and monkeys.
  • No acute CNS or GI effects observed in rats.
  • Shows weak activity against hERG (IC50, 5.8 μM).
  • Inhibits chemotactic response to MIP-1α.
  • Suppresses RANTES-induced signaling.
  • Potently suppresses all tested viral isolates.
Catalog No. 50-003-84999 Supplier Medchemexpress LLC Supplier No. HY17377100MG
Vicriviroc maleate is a potent, selective, oral bioavailable and CNS penetrated antagonist of CCR5, with a Ki of 2.5 nM. It also inhibits HIV-1 in PBMC cells, with IC90s of 3.3 nM (JrFL), 2.8 nM (ADA-M), 1.8 nM (301657), 4.9 nM (JV1083) and 10 nM (RU 570).
  • Potent, selective, and oral bioavailable inhibitor of CCR5.
  • Inhibits HIV-1 in PBMC cells.
  • Demonstrates a mean IC50 and IC90 of 0.45 nM and 4 nM for a panel of HIV isolates.
  • Exhibits good oral availability in rats and monkeys.
  • No acute CNS or GI effects observed in rats.
  • Shows weak activity against hERG (IC50, 5.8 μM).
  • Inhibits chemotactic response to MIP-1α.
  • Suppresses RANTES-induced signaling.
  • Potently suppresses all tested viral isolates.