Learn More
Medchemexpress LLC Vicriviroc maleate | 599179-03-0 | MFCD09833218 | 99.9% | 649.70 | 1 ML
SDP
Catalog No. 5000385533
Encompass_Preferred
Vicriviroc maleate is a potent, selective, and orally bioavailable CCR5 antagonist. It is CNS penetrated with a Ki of 2.5 nM. This compound effectively inhibits HIV-1 in PBMC cells across various strains.
  • Potent and selective CCR5 antagonist
  • Oral bioavailable and CNS penetrated
  • Inhibits HIV-1 in PBMC cells
  • Active against multiple HIV-1 strains
Catalog No. 50-003-85533 Supplier Medchemexpress LLC Supplier No. HY1737710MM/1ML
Vicriviroc maleate is a potent, selective, and orally bioavailable CCR5 antagonist. It is CNS penetrated with a Ki of 2.5 nM. This compound effectively inhibits HIV-1 in PBMC cells across various strains.
  • Potent and selective CCR5 antagonist
  • Oral bioavailable and CNS penetrated
  • Inhibits HIV-1 in PBMC cells
  • Active against multiple HIV-1 strains