Learn More
Medchemexpress LLC Vicriviroc maleate | 599179-03-0 | MFCD09833218 | 99.9% | 649.70 g/mol | C32H42F3N5O6 | 10 MG
SDP
Catalog No. 5000297924
Encompass_Preferred
Vicriviroc maleate is the maleate salt of vicriviroc, a potent and selective CCR5 antagonist used as a research compound. It is orally bioavailable and CNS penetrant, with a reported Ki of approximately 2.5 nM and low-nanomolar antiviral activity in cell assays. Supplied as a high-purity white to off-white solid for laboratory research.
  • Potent CCR5 antagonist (Ki ≈ 2.5 nM).
  • Inhibits HIV-1 entry in cell assays at low nanomolar concentrations.
  • High purity suitable for research use (>99.9%).
  • White to off-white solid form.
  • Stable when stored sealed at 4°C; protect from moisture.
  • Commonly used in antiviral and chemokine receptor research.
Catalog No. 50-002-97924 Supplier Medchemexpress LLC Supplier No. HY1737710MG
Vicriviroc maleate is the maleate salt of vicriviroc, a potent and selective CCR5 antagonist used as a research compound. It is orally bioavailable and CNS penetrant, with a reported Ki of approximately 2.5 nM and low-nanomolar antiviral activity in cell assays. Supplied as a high-purity white to off-white solid for laboratory research.
  • Potent CCR5 antagonist (Ki ≈ 2.5 nM).
  • Inhibits HIV-1 entry in cell assays at low nanomolar concentrations.
  • High purity suitable for research use (>99.9%).
  • White to off-white solid form.
  • Stable when stored sealed at 4°C; protect from moisture.
  • Commonly used in antiviral and chemokine receptor research.