Learn More
TARGETMOL CHEMICALS INC VICRIVIROC MALEATE 5MG
SDP
Catalog No. 502580266
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vicriviroc maleate (SCH-417690 (maleate))(Sch-417690) is a piperazine-based CCR5 receptor antagonist with activity against human immunodeficiency virus. The IC50 value is 0.91 nM in clinical development for the treatment of HIV-1. purity: 97%
Catalog No. 50-258-0266 Supplier TARGETMOL CHEMICALS INC Supplier No. T34355MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Vicriviroc maleate (SCH-417690 (maleate))(Sch-417690) is a piperazine-based CCR5 receptor antagonist with activity against human immunodeficiency virus. The IC50 value is 0.91 nM in clinical development for the treatment of HIV-1. purity: 97%