Learn More
Med Vet International Ethicon Coated Vicryl Polyglactin 910 Suture, Violet, 6-0, 1X8" S-14 Double Armed, Absorbable, 8mm, 12/box
Catalog No. 503045040
Encompass_Preferred
The Ethicon J578G suture is a high-quality absorbable coated vicryl surgical suture. It measures 8" in length, has a size of 6-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a S-14 double armed reverse cutting spatula edge needle and has 12 sutures in a box.
Catalog No. 50-304-5040 Supplier Med Vet International Supplier No. J578G
The Ethicon J578G suture is a high-quality absorbable coated vicryl surgical suture. It measures 8" in length, has a size of 6-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a S-14 double armed reverse cutting spatula edge needle and has 12 sutures in a box.