Learn More
Abcam Victoria blue B, Nuclear stain, 25G
Catalog No. 502704767
Encompass_Preferred
MW 506.1 g/mol. Used as a nuclear stain in cytology/histochemical applications. Can be used in the quantitative determination of phospholipids.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4767 Supplier Abcam Supplier No. AB14627425G
MW 506.1 g/mol. Used as a nuclear stain in cytology/histochemical applications. Can be used in the quantitative determination of phospholipids.
The product is subject to the following: Abcam Restricted Use Statement