Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Victoria Pure Blue BO, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013653 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3653 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3653 Supplier MilliporeSigma Supelco Supplier No. 7677325MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 2390-60-5
Physical Form Neat
Linear Formula C33H40 ClN3
MDL Number MFCD00011877
Synonym Basic Blue 7
Type Analytical Standard
Quantity 25 mg
Formula Weight 514.14
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.