Learn More
Medchemexpress LLC Vidarabine monohydrate | 24356-66-9 | MFCD00150980 | 1 ML
SDP
Catalog No. 5000418821
Encompass_Preferred
Vidarabine monohydrate is an adenine arabinoside and a potent antiviral agent. It effectively targets herpes simplex viruses (HSV) and varicella zoster viruses.
  • Adenine arabinoside
  • Antiviral agent
  • Active against herpes simplex viruses (HSV)
  • Active against varicella zoster viruses
Catalog No. 50-004-18821 Supplier Medchemexpress LLC Supplier No. HYN666610MM/1ML
Vidarabine monohydrate is an adenine arabinoside and a potent antiviral agent. It effectively targets herpes simplex viruses (HSV) and varicella zoster viruses.
  • Adenine arabinoside
  • Antiviral agent
  • Active against herpes simplex viruses (HSV)
  • Active against varicella zoster viruses