Learn More
Medchemexpress LLC HY-N6666 50mg Medchemexpress, Vidarabine (monohydrate) CAS:24356-66-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN666650MG
Encompass_Preferred
Medchemexpress, HY-N6666 50mg Vidarabine (monohydrate) CAS:24356-66-9 Vidarabine monohydrate is an adenine arabinoside. Vidarabine monohydrate an antiviral drug which is active against herpes simplex viruses (HSV) and varicella zoster viruses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2954