Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vidarabine phosphate | 29984-33-6 | 99.4% | 25 MG
SDP

Catalog No. 5000450564
Encompass_Preferred

Vidarabine phosphate, also known as Ara-AMP, is an antiviral agent primarily used to inhibit chronic HBV infection. This compound has also shown effectiveness against herpes simplex and varicella zoster viruses, making it a versatile agent in antiviral research.

  • Antiviral properties
  • Inhibits chronic HBV infection
  • Effective against herpes simplex
  • Effective against varicella zoster viruses
  • White to off-white solid appearance
  • Molecular weight of 347.22
  • Chemical formula C10H14N5O7P
  • High purity of 99.4%

Catalog No. 50-004-50564 Supplier Medchemexpress LLC Supplier No. 25MGHYB0277A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vidarabine phosphate, also known as Ara-AMP, is an antiviral agent primarily used to inhibit chronic HBV infection. This compound has also shown effectiveness against herpes simplex and varicella zoster viruses, making it a versatile agent in antiviral research.

  • Antiviral properties
  • Inhibits chronic HBV infection
  • Effective against herpes simplex
  • Effective against varicella zoster viruses
  • White to off-white solid appearance
  • Molecular weight of 347.22
  • Chemical formula C10H14N5O7P
  • High purity of 99.4%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.