Learn More
Medchemexpress LLC Vidarabine phosphate | 29984-33-6 | MFCD00069723 | 98.7% | 347.22 g/mol | C10H14N5O7P | 10 MG
SDP
Catalog No. 5000297403
Encompass_Preferred
Vidarabine phosphate is the 5'-monophosphate form of the antiviral nucleoside vidarabine (Ara-A). It is used as a research reagent in antiviral and biochemical assays and has reported activity against herpes simplex virus, varicella zoster virus, and chronic hepatitis B virus.
  • Antiviral nucleoside monophosphate active against HSV, VZV, and HBV.
  • Supplied as a solid powder and available as stock solution for biochemical assays.
  • High purity suitable for research applications.
  • Stable when stored sealed and protected from moisture; stock solutions require low-temperature storage.
  • Useful as a biochemical standard or reagent in antiviral mechanism studies.
Catalog No. 50-002-97403 Supplier Medchemexpress LLC Supplier No. HYB0277A10MG
Vidarabine phosphate is the 5'-monophosphate form of the antiviral nucleoside vidarabine (Ara-A). It is used as a research reagent in antiviral and biochemical assays and has reported activity against herpes simplex virus, varicella zoster virus, and chronic hepatitis B virus.
  • Antiviral nucleoside monophosphate active against HSV, VZV, and HBV.
  • Supplied as a solid powder and available as stock solution for biochemical assays.
  • High purity suitable for research applications.
  • Stable when stored sealed and protected from moisture; stock solutions require low-temperature storage.
  • Useful as a biochemical standard or reagent in antiviral mechanism studies.