Learn More
Cole-Parmer Meiji Techno MA151/5N Microscope Eyepiece to Video/Camera Adapter; for 48403-10, 48402-00/-02
Catalog No. 50631623 Shop All Cole-Parmer Products
Encompass
Use adapters to connect an eyepiece or trinocular microscope to a video camera with a "C" mount. Adapter assembly 03907-50 easily slips over the 28-mm eyepiece of most microscopes.
Catalog No. 50-631-623 Supplier Cole-Parmer Supplier No. 4840272
Use adapters to connect an eyepiece or trinocular microscope to a video camera with a "C" mount. Adapter assembly 03907-50 easily slips over the 28-mm eyepiece of most microscopes.