Learn More
Apexbio Technology LLC Vidofludimus, 10mg. Cas: 717824-30-1 MFCD: MFCD18633262. DHODH inhibitor
SDP
Supplier:  Apexbio Technology LLC B466910
Encompass_Preferred
Vidofludimus (4SC-101, SC12267) is a novel small molecule inhibitor of dihydroorotate dehydrogenase (DHODH) with an IC50 value of 134nmol/L for human DHODH [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5355
 Restricted Item