Learn More
Cayman Chemical VIDOFLUDIMUS 10MG
SDP
Catalog No. 502979473
Encompass_Preferred
An immunosuppressive drug that inhibits DHODH (IC50s = 0.134, 1.29, and 10.6 µM for human, rat, and mouse isoforms, respectively); inhibits the proliferation of T cells and B cells and the secretion of IL-17 (IC50s = 12.9, 3.7, and 6.0 µM, respectively, in human cells)
Catalog No. 50-297-9473 Supplier Cayman Chemical Supplier No. 1837710 MG
An immunosuppressive drug that inhibits DHODH (IC50s = 0.134, 1.29, and 10.6 µM for human, rat, and mouse isoforms, respectively); inhibits the proliferation of T cells and B cells and the secretion of IL-17 (IC50s = 12.9, 3.7, and 6.0 µM, respectively, in human cells)