Learn More
Bioworld VIEWIP ANTI-HA RED SEPAROPO
Catalog No. 50197941
Encompass
VIEWIP ANTI-HA RED SEPAROPORE (AGAROSE): Red-Colored Affinity matrix consists of affinity purified anti-HA mouse monoclonal antibody (Clone HA-7) covalently attached to beaded agarose. The HA tag corresponds to amino acid residues 98-106...
Catalog No. 50-197-941 Supplier Bioworld Supplier No. 201811151
VIEWIP ANTI-HA RED SEPAROPORE (AGAROSE): Red-Colored Affinity matrix consists of affinity purified anti-HA mouse monoclonal antibody (Clone HA-7) covalently attached to beaded agarose. The HA tag corresponds to amino acid residues 98-106...