Learn More
Bioworld viewIP Calmodulin RedSeparopore (Agarose) 4B-CL
Supplier:  Bioworld 201811041
Encompass
Applications: Red-coloredagarose matrix contains immobilizedbovine calmodulinfor convenient affinity capture (pull-down) target molecules. viewIP™Calmodulin Red-Separopore 4B-CL greatly reduce the risk of pellet loss. Comparable performance to non-colored affinity gels, but allows the user to easily differentiate pellet from supernatant. This results in greater consistencyand more accurate data due to less sample losses.viewIP™Calmodulin Red-Separopore 4B-CL is used for convenient affinity purification of calmodulin-binding proteins including calmodulin-regulated proteins in eukaryotic cells and recombinant proteins containing calmodulin-binding peptide fusion tag.
Catalog No. 50-197-925