Learn More
Bioworld VIEWIP GLUTATHIONE RED SEPA
Supplier:  Bioworld 201811161
Encompass
VIEWIP GLUTATHIONE RED SEPAROPORE (AGAROSE): Red-Colored Affinity matrix consists glutathione (reduced) covalently attached to red beaded agarose 4B (Separopore) via epoxy activation. Used for affinity capture, molecular pull-down, or IP...
Catalog No. 50-197-942