Learn More
Bioworld VIEWIP PROTEIN-A BLUE SEPAR
Supplier:  Bioworld 201811052
Encompass
VIEWIP PROTEIN-A BLUE SEPAROPORE (AGAROSE) 4B-CL: viewIP Protein-ABlue Separopore 4B-CL Blue-Colored Affinity matrixhas immobilized Recombinant Protein-A. Convenient for quantitative capturing of antigen/antibody complexes in immunoprec...
Catalog No. 50-197-928