Learn More
Bioworld VIEWIP PROTEIN-A BLUE SEPAR
Supplier: Bioworld 201811051
VIEWIP PROTEIN-A BLUE SEPAROPORE (AGAROSE) 4B-CL: viewIP Protein-ABlue Separopore 4B-CL Blue-Colored Affinity matrixhas immobilized Recombinant Protein-A. Convenient for quantitative capturing of antigen/antibody complexes in immunoprec...