Learn More
Bioworld VIEWIP PROTEIN-A GREEN SEPA
Supplier:  Bioworld 201811082
Encompass
VIEWIP PROTEIN-A GREEN SEPAROPORE (AGAROSE) 4B-CL: viewIP Protein-AGreen Separopore 4B-CL Geen-Colored Affinity matrixhas immobilized Recombinant Protein-A. Convenient for quantitative capturing of antigen/antibody complexes in immunopr...
Catalog No. 50-197-932