Learn More
Bioworld VIEWIP PROTEIN-A RED SEPARO
Supplier:  Bioworld 201810411
Encompass
VIEWIP PROTEIN-A RED SEPAROPORE (AGAROSE) 4B-CL: viewIP Protein-A Red Separopore 4B-CL Red-Colored Affinity matrixhas immobilized Recombinant Protein-A. Convenient for quantitative capturing of antigen/antibody complexes in immunoprecip...
Catalog No. 50-197-768