Learn More
Bioworld VIEWIP PROTEIN G RED SEPARO
Catalog No. 50197944
Encompass
VIEWIP PROTEIN G RED SEPAROPORE (AGAROSE) 4B-CL: Red-Colored Affinity matrix consists of immobilized protein-G (from Streptococcus sp.) covalently attached to red beaded agarose (Separopore 4B). Convenient for small-scale affinity captur...
Catalog No. 50-197-944 Supplier Bioworld Supplier No. 201811181
VIEWIP PROTEIN G RED SEPAROPORE (AGAROSE) 4B-CL: Red-Colored Affinity matrix consists of immobilized protein-G (from Streptococcus sp.) covalently attached to red beaded agarose (Separopore 4B). Convenient for small-scale affinity captur...