Learn More
Bioworld VIEWIP STREPTAVIDIN GREEN S
Catalog No. 50197933
Encompass
VIEWIP STREPTAVIDIN GREEN SEPAROPORE (AGAROSE) 4B-CL: viewIP StreptavidinGreen Separopore 4B-CL Green-Coloredagarose matrix contains immobilized Streptavidin for convenient affinity capture (pull-down) of biotinylated target molecules, ...
Catalog No. 50-197-933 Supplier Bioworld Supplier No. 201811091
VIEWIP STREPTAVIDIN GREEN SEPAROPORE (AGAROSE) 4B-CL: viewIP StreptavidinGreen Separopore 4B-CL Green-Coloredagarose matrix contains immobilized Streptavidin for convenient affinity capture (pull-down) of biotinylated target molecules, ...