Learn More
Bioworld VIEWIP STREPTAVIDIN RED SEP
Supplier:  Bioworld 201811031
Encompass
VIEWIP STREPTAVIDIN RED SEPAROPORE (AGAROSE) 4B-CL: viewIP Streptavidin Red Separopore 4B-CL Red-colored agarose matrix contains immobilized Streptavidin for convenient affinity capture (pull-down) of biotinylated target molecules, such...
Catalog No. 50-197-923