Learn More
Revvity Health Sciences Inc ViewPlate-384 Black, Optically Clear Bottom, Tissue Culture Treated, Sterile, 384-Well with Lid, Case of 40
Catalog No. 509052489
Encompass
ViewPlate-384 Black, Optically Clear Bottom, Tissue Culture Treated, Sterile, 384-Well with Lid, Case of 40
Catalog No. 50-905-2489 Supplier Revvity Health Sciences Inc Supplier No. 6007460
ViewPlate-384 Black, Optically Clear Bottom, Tissue Culture Treated, Sterile, 384-Well with Lid, Case of 40