Learn More
Revvity Health Sciences Inc ViewPlate-384 Collagen Coated, Black, Optically Clear Bottom, Tissue Culture Treated, Sterile, 384-Well with Lid, Case of 40
Catalog No. 502105675
Encompass
ViewPlate-384 Collagen Coated, Black, Optically Clear Bottom, Tissue Culture Treated, Sterile, 384-Well with Lid, Case of 40
Catalog No. 50-210-5675 Supplier Revvity Health Sciences Inc Supplier No. 6007810
ViewPlate-384 Collagen Coated, Black, Optically Clear Bottom, Tissue Culture Treated, Sterile, 384-Well with Lid, Case of 40