Learn More
Revvity Health Sciences Inc ViewPlate-384 PDL Coated, Black, Optically Clear Bottom, Tissue Culture Treated, Sterile, 384-Well with Lid, Case of 160
Catalog No. 502105679
Encompass
ViewPlate-384 PDL Coated, Black, Optically Clear Bottom, Tissue Culture Treated, Sterile, 384-Well with Lid, Case of 160
Catalog No. 50-210-5679 Supplier Revvity Health Sciences Inc Supplier No. 6007718
ViewPlate-384 PDL Coated, Black, Optically Clear Bottom, Tissue Culture Treated, Sterile, 384-Well with Lid, Case of 160