Learn More
Revvity Health Sciences Inc ViewPlate-96 Collagen Coated, Black, Optically Clear Bottom, 96-Well with Lid, Case of 40
Catalog No. 502105844
Encompass
ViewPlate-96 Collagen Coated, Black, Optically Clear Bottom, 96-Well with Lid, Case of 40
Catalog No. 50-210-5844 Supplier Revvity Health Sciences Inc Supplier No. 6005810
ViewPlate-96 Collagen Coated, Black, Optically Clear Bottom, 96-Well with Lid, Case of 40