Learn More
Thermo Scientific™ ViewRNA, 1440 μL
Description
ViewRNA, 1440 μL
Specifications
Specifications
| Product Type | RNA In Situ Hybridization Reagent |
| Quantity | 1440 μL |
Frequently Asked Questions (FAQs)
Catalog ViewRNA probe sets can be ordered using a catalog number corresponding to the aliquot size selected:
- Small: 120 µL (Cat. No. VX-06)
- Medium: 440 µL (Cat. No. VX-01)
- Large: 1.44 mL (Cat. No. VX-02)
VX- is used for catalog ViewRNA probe sets and CVX- is used for custom-designed ViewRNA probe sets. To add ViewRNA probe sets to the cart, use the Quick Order portal and add the combination of the catalog number and the associated Assay ID. Alternatively, you may request a quote from your Sales specialist or by contacting Technical Support at cellanalysis.support@thermofisher.com.
ViewRNA probe sets are available in 3 sizes:
- Small: 120 µL
- Medium: 440 µL
- Large: 1.44 mL
ViewRNA probe sets are provided as concentrated stock solutions that need to be diluted in the assay protocol. The dilution factors, as well as the volume per sample vary between assays, based on the type of vessel used. Hence the number of sustained assays will also vary. Cell assays typically require 1:100 dilution while tissue assays can use 1:40 or 1:20 dilution.
If for example, the protocol calls for 1:100 dilution and you purchased the Medium aliquot, this will make 44 mL of the 1X probe set solution: 440 µL x 100 = 44,000 µl.
If using a standard protocol that requires 405 µL of 1X probe set per slide (or sample), the Medium aliquot will sustain 108 assays in total: 44,000 µL/ 405 µL = 108 assays.
PrimeFlow probe sets can be used for ViewRNA assays and vice versa but note that the 'Type' designation is not equivalent. Please refer to the table below for Type-fluorophore assignments per each assay.

The design strategy for a ViewRNA probe set involves performing a BLAST of the target gene sequence to identify a unique, non-repetitive region for complete specificity. The target region of a 'standard' ViewRNA probe set is ~800 bp long, helping ensure maximum strength of the probe set. ViewRNA probes are branched DNA probe pairs, called ZZ pairs with each oligo in the pair being ~20 bp long. The left and right ZZ oligos have to align adjacent to each other, otherwise, signal amplification will not occur. This helps ensure the most specific detection. The ViewRNA probe set can consist of anywhere between 1-20 oligo pairs. Even a single oligo pair can help provide resolution down to a single transcript.
The ViewRNA probe sets come with 12-month warranty, if stored as indicated.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
