Learn More
Invitrogen™ ViewRNA™, 1440 μL
Catalog No. CVX02
Encompass_Preferred
Change view
Click to view available options
Quantity:
1440 μL
Catalog No. Quantity
CVX02 1440 μL
1 options
Catalog No. CVX02 Supplier Invitrogen™ Supplier No. CVX02
Add to Cart
Add to Cart
This is a customizable probe set for the ViewRNA ISH Cell, ViewRNA ISH Tissue, and ViewRNA Cell Plus assays.
This is a customizable probe set for the ViewRNA ISH Cell, ViewRNA ISH Tissue, and ViewRNA Cell Plus assays.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage ViewRNA ISH probe set, 1.44 mL
Store at -20°C.
Quantity 1440 μL
Product Line ViewRNA
How can I order custom ViewRNA probe sets?
Please use the Custom Branched DNA Probe Set Tool (https://www.thermofisher.com/order/custom-oligo/brancheddna) to order custom ViewRNA probe sets.