Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ViewRNA™ Cell Plus Cytospin Module Kit

Catalog No. 881900211
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-190-0211 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-190-0211 Supplier Invitrogen™ Supplier No. 881900211
Add to Cart
Add to Cart

The ViewRNA™ Cell Plus Cytospin Module Kit is for use with the ViewRNA™ Cell Plus Assay kit (Cat. No. 88-19000).

The ViewRNA™ Cell Plus Cytospin Module Kit is for use with the ViewRNA™ Cell Plus Assay kit (Cat. No. 88-19000). These buffers can be used to perform the assay when using cytospin slides with cells.

Please refer to the ViewRNA™ Cell Plus Assay protocol on the product data sheet for instructions for use.

Reported Application

  • Microscopy, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
For Use With (Application) Microscopy, Immunocytochemistry
For Use With (Equipment) ViewRNA™ Cell Plus Assay Kit (Cat. No. 88-19000)
Product Type Cytospin Module Kit
Quantity 1 Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.